Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Dimethylamino)benzaldehyde Solution 10 g/L in Isopropanol, For TLC Derivatization, MilliporeSigma™ Supelco™

Catalog No. 111018553 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8553 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8553 Supplier MilliporeSigma Supelco Supplier No. 02560500ML
Add to Cart
Add to Cart

Specifications

CAS 100-10-7
Linear Formula C9H11NO
MDL Number MFCD00003381
Quantity 500 mL
Formula Weight 149.19
Grade For TLC derivatization
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.