Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Dimethylamino)benzenethiol 98.0+%, TCI America™
SDP

Catalog No. D40471G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D40471G Supplier TCI America Supplier No. D40471G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4946-22-9
Molecular Formula C8H11NS
Molecular Weight (g/mol) 153.243
MDL Number MFCD01658123
InChI Key PQSBRHXGVPVYFJ-UHFFFAOYSA-N
Synonym 4-(Dimethylamino)thiophenol, 4-Mercapto-N,N-dimethylaniline
PubChem CID 21061
IUPAC Name 4-(dimethylamino)benzenethiol
SMILES CN(C)C1=CC=C(C=C1)S

Specifications

Melting Point 33°C
Color Yellow
Boiling Point 260°C
Quantity 1 g
Formula Weight 153.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Dimethylamino)benzenethiol
RTECSNumber DC1575000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.