Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Dimethylamino)butyraldehyde Dimethyl Acetal 98.0+%, TCI America™
SDP

Catalog No. D35355G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35355G 5 g
D353525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35355G Supplier TCI America Supplier No. D35355G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19718-92-4
Molecular Formula C8H19NO2
Molecular Weight (g/mol) 161.245
MDL Number MFCD06411126
InChI Key WDZKKBDOGYBYBG-UHFFFAOYSA-N
Synonym 4,4-Dimethoxy-N,N-dimethylbutylamine
PubChem CID 10374734
IUPAC Name 4,4-dimethoxy-N,N-dimethylbutan-1-amine
SMILES CN(C)CCCC(OC)OC

Specifications

Color Yellow
Boiling Point 54°C
Quantity 5 g
Formula Weight 161.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(Dimethylamino)butyraldehyde Dimethyl Acetal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.