Learn More
Sigma Aldrich Fine Chemicals Biosciences 4-(Dimethylamino)cinnamaldehyde chromogenic reagent for indoles and flavanols | 6203-18-5 | MFCD00007002 | 10G
Catalog No. 501840783
Encompass
4-(Dimethylamino)cinnamaldehyde chromogenic reagent for indoles and flavanols | Purity: >=98% (HPLC) | Mol Wt: 175.23 | 6203-18-5 | MFCD00007002 | 10G
Catalog No. 501840783 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D450610G
4-(Dimethylamino)cinnamaldehyde chromogenic reagent for indoles and flavanols | Purity: >=98% (HPLC) | Mol Wt: 175.23 | 6203-18-5 | MFCD00007002 | 10G