Learn More
Medchemexpress LLC 4-(Dimethylamino)cinnamaldehyde | 6203-18-5 | MFCD00007002 | 99.8% | 175.23 g/mol | C11H13NO | 10 G
SDP
Catalog No. 5000303241
Encompass_Preferred
4-(Dimethylamino)cinnamaldehyde is an organic reagent commonly used in biochemical assays to detect indole-derived products and to quantify proanthocyanidins in plant extracts. It is supplied as a yellow-orange solid with high analytical purity and documented storage and solubility properties for reliable laboratory use.
  • Reagent for detecting indole products in enzymatic assays.
  • Applicable for quantifying proanthocyanidins in plant extracts.
  • High purity suitable for analytical and assay workflows.
  • Yellow-orange solid with established storage conditions for stability.
  • Readily soluble in DMSO (100 mg/mL) with gentle warming for in vitro use.
Catalog No. 50-003-03241 Supplier Medchemexpress LLC Supplier No. HYW01672910G
4-(Dimethylamino)cinnamaldehyde is an organic reagent commonly used in biochemical assays to detect indole-derived products and to quantify proanthocyanidins in plant extracts. It is supplied as a yellow-orange solid with high analytical purity and documented storage and solubility properties for reliable laboratory use.
  • Reagent for detecting indole products in enzymatic assays.
  • Applicable for quantifying proanthocyanidins in plant extracts.
  • High purity suitable for analytical and assay workflows.
  • Yellow-orange solid with established storage conditions for stability.
  • Readily soluble in DMSO (100 mg/mL) with gentle warming for in vitro use.