Learn More
Medchemexpress LLC 4-(dimethylamino)cinnamaldehyde | 6203-18-5 | MFCD00007002 | 99.8% | 175.23 g/mol | C11H13NO | 50 G
SDP
Catalog No. 5000303334
Encompass_Preferred
4-(Dimethylamino)cinnamaldehyde is a biochemical assay reagent used as a chromogenic reagent for detecting indole-containing compounds and for electrophoresis gel preparation. It is typically supplied as a yellow-to-orange powder; CAS 6203-18-5, molecular formula C11H13NO, molecular weight 175.23 g/mol, with Certificate of Analysis reporting 99.82% (HPLC).
  • Detects indole products in biochemical assays.
  • Provides chromogenic color development for colorimetric detection.
  • Suitable for electrophoresis gel preparation.
  • High purity supports reproducible analytical results.
  • Solid powder form for convenient storage and handling.
  • Molecular formula C11H13NO; molecular weight 175.23 g/mol.
Catalog No. 50-003-03334 Supplier Medchemexpress LLC Supplier No. HYW01672950G
4-(Dimethylamino)cinnamaldehyde is a biochemical assay reagent used as a chromogenic reagent for detecting indole-containing compounds and for electrophoresis gel preparation. It is typically supplied as a yellow-to-orange powder; CAS 6203-18-5, molecular formula C11H13NO, molecular weight 175.23 g/mol, with Certificate of Analysis reporting 99.82% (HPLC).
  • Detects indole products in biochemical assays.
  • Provides chromogenic color development for colorimetric detection.
  • Suitable for electrophoresis gel preparation.
  • High purity supports reproducible analytical results.
  • Solid powder form for convenient storage and handling.
  • Molecular formula C11H13NO; molecular weight 175.23 g/mol.