Learn More
Medchemexpress LLC 4-(dimethylamino)phenacyl bromide-13C2,d6 | MFCD06409221 | 98.1% | 250.13 g/mol | C8 13C2 H6 D6 Br N O | 100 MG
SDP
Catalog No. 5000282620
Encompass_Preferred
4-(Dimethylamino)phenacyl bromide-13C2,d6 is a stable isotope-labeled analog of 4-(dimethylamino)phenacyl bromide intended for research use. It functions as an internal standard and tracer in analytical workflows, supporting accurate quantification, metabolite tracking, and method validation in mass spectrometry and NMR applications.
  • Isotopically labeled with two 13C atoms and six deuterium atoms.
  • High reported purity of 98.14%.
  • Suitable as an internal standard for LC-MS, GC-MS, and NMR applications.
  • Helps improve quantitation accuracy and correct for matrix effects.
  • Available in multiple small-scale quantities for analytical workflows.
Catalog No. 50-002-82620 Supplier Medchemexpress LLC Supplier No. HYW128417S100MG
4-(Dimethylamino)phenacyl bromide-13C2,d6 is a stable isotope-labeled analog of 4-(dimethylamino)phenacyl bromide intended for research use. It functions as an internal standard and tracer in analytical workflows, supporting accurate quantification, metabolite tracking, and method validation in mass spectrometry and NMR applications.
  • Isotopically labeled with two 13C atoms and six deuterium atoms.
  • High reported purity of 98.14%.
  • Suitable as an internal standard for LC-MS, GC-MS, and NMR applications.
  • Helps improve quantitation accuracy and correct for matrix effects.
  • Available in multiple small-scale quantities for analytical workflows.