Learn More
Medchemexpress LLC 4-(Dimethylamino)phenacyl bromide-d6 | 37904-72-6 | MFCD06409221 | 98.8% | 248.15 g/mol | C10H6D6BrNO | 10 MG
SDP
Catalog No. 5000282751
Encompass_Preferred
4-(Dimethylamino)phenacyl bromide-d6 is a deuterium-labeled isotopologue intended as an internal standard and tracer for quantitative analytical methods. Supplied as a white to off-white solid, it provides a mass-shifted reference for precise measurement in complex matrices and is intended for research use only.
  • Deuterium-labeled (d6) internal standard for analytical quantitation.
  • Suitable for NMR, GC-MS, and LC-MS as a tracer or internal standard.
  • Molecular formula C10H6D6BrNO; molecular weight 248.15 g/mol.
  • High purity for analytical use (98.8%).
  • Stable powder with recommended storage: powder -20°C (3 years) or 4°C (2 years).
  • White to off-white solid supplied in 10 MG packaging.
  • For research use only; not for human or animal therapeutic use.
Catalog No. 50-002-82751 Supplier Medchemexpress LLC Supplier No. HYW128417S110MG
4-(Dimethylamino)phenacyl bromide-d6 is a deuterium-labeled isotopologue intended as an internal standard and tracer for quantitative analytical methods. Supplied as a white to off-white solid, it provides a mass-shifted reference for precise measurement in complex matrices and is intended for research use only.
  • Deuterium-labeled (d6) internal standard for analytical quantitation.
  • Suitable for NMR, GC-MS, and LC-MS as a tracer or internal standard.
  • Molecular formula C10H6D6BrNO; molecular weight 248.15 g/mol.
  • High purity for analytical use (98.8%).
  • Stable powder with recommended storage: powder -20°C (3 years) or 4°C (2 years).
  • White to off-white solid supplied in 10 MG packaging.
  • For research use only; not for human or animal therapeutic use.