Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Dimethylamino)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. D44285G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4428-5G 5 g
D44281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4428-5G Supplier TCI America Supplier No. D44285G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material 4-Dimethylamino Phenylboronic Acid
CAS 28611-39-4
Melting Point 227°C
Color White-Yellow
Molecular Formula C8H12BNO2
MDL Number MFCD01074642
Quantity 5 g
Synonym 4-dimethylamino phenylboronic acid, 4-dimethylamino phenyl boronic acid, 4-n,n-dimethylamino phenylboronic acid, 4-dimethylamino benzeneboronic acid, 4-dimethylaminophenylboronic acid
InChI Key RIIPFHVHLXPMHQ-UHFFFAOYSA-N
SMILES B(C1=CC=C(C=C1)N(C)C)(O)O
IUPAC Name [4-(dimethylamino)phenyl]boronic acid
Molecular Weight (g/mol) 164.999
PubChem CID 2734344
Formula Weight 165.00
Percent Purity 97.0 to 113.0%
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.