Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Dimethylamino)phenyldiphenylphosphine 95.0+%, TCI America™
SDP

Catalog No. D24785G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24785G 5 g
D24781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24785G Supplier TCI America Supplier No. D24785G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 739-58-2
Molecular Formula C20H20NP
Molecular Weight (g/mol) 305.36
MDL Number MFCD00192068
InChI Key GOEGBJDTWXTPHP-UHFFFAOYSA-N
Synonym 4-dimethylamino phenyldiphenylphosphine, 4-diphenylphosphino-n,n-dimethylaniline, 4-diphenylphosphanyl-n,n-dimethylaniline, 4-dimethylamino triphenylphosphine, benzenamine, 4-diphenylphosphino-n,n-dimethyl, 4-dimethylaminophenyl diphenylphosphine, 4-diphenylphosphino-n,n-dimethyl-benzenamine, acmc-209ot0, ksc495a7n, p-dimethylaminophenyl-diphenyl-phosphine
PubChem CID 3333592
IUPAC Name 4-(diphenylphosphanyl)-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 152°C
Color White
Quantity 5 g
Formula Weight 305.36
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Dimethylamino)phenyldiphenylphosphine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.