Learn More
Medchemexpress LLC 4-(dimethylamino)pyridine | 1122-58-3 | MFCD00006418 | 122.17 g/mol | C7H10N2 | 1000g
SDP
Catalog No. 5000267125
Encompass_Preferred
4-Dimethylaminopyridine is an acyl transfer catalyst 4-Dimethylaminopyridine is used for labeling and analysis of glycoproteins on the surface of living cells 4-Dimethylaminopyridine reduces dyskinesia attacks[1][2]
Catalog No. 50-002-67125 Supplier Medchemexpress LLC Supplier No. HYY10891000G
4-Dimethylaminopyridine is an acyl transfer catalyst 4-Dimethylaminopyridine is used for labeling and analysis of glycoproteins on the surface of living cells 4-Dimethylaminopyridine reduces dyskinesia attacks[1][2]