Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-(Dimethylamino)Pyridine, Polymer-Bound

Catalog No. 394105G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-410-5G 5 g
39-410-25G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 39-410-5G Supplier Sigma Aldrich Supplier No. 394105G
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Polymer-bound 4-(dimethylamino)pyridine is used for improved derivatization of chiral and achiral aliphatic amines, amino alcohols and amino acids.Polymer-bound 4-(dimethylamino)pyridine is used as a supported base in the Michael addition and nitrous acid elimination process step involved in the synthesis of thiophene-2-carboxylates using +a-nitroacrylates as starting materials, under a general and efficient continuous flow-based protocol. It also serves as a supported catalyst in the preparation of -d-nitro esters starting from cyclic 2-nitro ketones under solid heterogeneous catalysis.(5)Polymer supported, recyclable acylation catalyst

Specifications

CAS 82942-26-5
Molecular Formula C8H8
Linear Formula C8H8
MDL Number MFCD04041220
Quantity 5 g
Molecular Weight (g/mol) 104.15 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.