Learn More
Cayman Chemical 4 DImethylamIno1methylpyr 50mg
SDP
Catalog No. 502933428
Encompass_Preferred
A pyridinium salt with anticholinesterase and antiproliferative activities; inhibits cholinesterase activity in rat brain homogenates (Kd = 33 μM); inhibits and yeast ChoK (IC50 = 17 μM); has antiproliferative activity against HT-29 colon cancer cells (IC50 = 2 μM)
Catalog No. 50-293-3428 Supplier Cayman Chemical Supplier No. 2586150 MG
A pyridinium salt with anticholinesterase and antiproliferative activities; inhibits cholinesterase activity in rat brain homogenates (Kd = 33 μM); inhibits and yeast ChoK (IC50 = 17 μM); has antiproliferative activity against HT-29 colon cancer cells (IC50 = 2 μM)