Learn More
AdipoGen 4-Dimethylaminocinnamaldehyde
SDP
Supplier:  AdipoGen CDXD0310G005
Encompass_Preferred
Chemical. CAS 6203-18-5. Formula C11H13NO. MW 175.2. Synthetic. Primarily used as a histological dye to detect indoles. Reagent for the determination of aromatic amines. It is used for the rapid identification of bacteria containing apotryptophanase and tryptophanase enzyme systems. It is also particularly useful for localization of proanthocyanidins compounds in plants, resulting in a blue staining. Has been developed for flavanoids in beer and is used in forensics.
Catalog No. 50-256-1130