Learn More
Accela Chembio Inc 4-(dimethylaminomethyl)aniline | 25g | 6406-74-2 | MFCD00035953 | 97+% | Shelf Life: 1260 Days | Light Sensitive/nitrogen Or Argon
Supplier:  Accela Chembio Inc SY00357125G
Encompass_Preferred
4-(dimethylaminomethyl)aniline | 25g | 6406-74-2 | MFCD00035953 | 97+% | Shelf Life: 1260 Days | Light Sensitive/nitrogen Or Argon
Catalog No. 50-353-785