Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Dimethylaminophenyl)di-tert-butylphosphine, TCI America™
SDP

Catalog No. D45315G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4531-5G 5 g
D45311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4531-5G Supplier TCI America Supplier No. D45315G
Add to Cart
Add to Cart

Specifications

CAS 932710-63-9
Melting Point 69°C
Color White-Yellow
Molecular Formula C16H28NP
Quantity 5 g
MDL Number MFCD09265102
Synonym 4-(Di-tert-butylphosphino)-N,N-dimethylaniline, APhos, A-taPhos, Amphos
InChI Key IQTHEAQKKVAXGV-UHFFFAOYSA-N
SMILES CC(C)(C)P(C1=CC=C(C=C1)N(C)C)C(C)(C)C
IUPAC Name 4-ditert-butylphosphanyl-N,N-dimethylaniline
Molecular Weight (g/mol) 265.381
PubChem CID 11714598
Formula Weight 265.38
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.