Learn More
Medchemexpress LLC 4-dimethylaminopyridine | 1122-58-3 | MFCD00006418 | 100.0% | 122.17 g/mol | C7H10N2 | 1mL
SDP
Catalog No. 5000268005
Encompass_Preferred
4-Dimethylaminopyridine is an acyl transfer catalyst 4-Dimethylaminopyridine is used for labeling and analysis of glycoproteins on the surface of living cells 4-Dimethylaminopyridine reduces dyskinesia attacks[1][2]
Catalog No. 50-002-68005 Supplier Medchemexpress LLC Supplier No. HYY108910MM/1ML
Add to Cart
Add to Cart
4-Dimethylaminopyridine is an acyl transfer catalyst 4-Dimethylaminopyridine is used for labeling and analysis of glycoproteins on the surface of living cells 4-Dimethylaminopyridine reduces dyskinesia attacks[1][2]