Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α,4-Dimethylbenzyl alcohol, 98%, Thermo Scientific™

Catalog No. AC259690500
Change view
Click to view available options
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC259690500 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC259690500 Supplier Thermo Scientific Chemicals Supplier No. 259690500

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 536-50-5
CAS Min % 97.5
CAS Max % 100.0
Density 0.98g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C9H12O
Refractive Index 1.5210 to 1.5230 (20°C, 589nm)
Quantity 50mL
MDL Number MFCD00016860
Merck Index 15, 3259
Synonym 1-4-methylphenyl ethanol, 1-p-tolyl ethanol, bilagen, galinex, norbilan, tomobil, methyl-p-tolylcarbinol, p-tolylmethylcarbinol, 1-4-tolyl ethanol, 1-p-tolyl-1-ethanol
InChI Key JESIHYIJKKUWIS-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)C(C)O
IUPAC Name 1-(4-methylphenyl)ethanol
Molecular Weight (g/mol) 136.194
PubChem CID 10817
Formula Weight 136.18
Percent Purity ≥97.5%
Chemical Name or Material α, 4-Dimethylbenzyl alcohol, 98%
Show More Show Less
EINECSNumber 208-637-8
RTECSNumber DO4565000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.