Learn More
Medchemexpress LLC 4-diphenylacetoxy-N-methylpiperidinium iodide | 1952-15-4 | MFCD00078564 | >98.0% | 451.34 g/mol | C21H26INO2 | 1 ML
SDP
Catalog No. 5000211398
Encompass_Preferred
4-DAMP (4-diphenylacetoxy-N-methylpiperidinium iodide) is a quaternary ammonium muscarinic receptor antagonist selective for M3 and M5 subtypes, supplied for research applications. It is available as a ready-to-use 10 mM solution in DMSO (1 mL) and in solid forms, and is accompanied by analytical documentation for quality assurance.
  • Ready-to-use 10 mM solution in DMSO for convenient dosing and preparation.
  • High purity suitable for research applications (>98% by HPLC).
  • Appropriate for pharmacological, receptor binding, and signaling studies.
  • Supplied with supporting analytical documents such as COA, HNMR, and RP-HPLC.
  • Molecular weight 451.3 g/mol; CAS 1952-15-4.
Catalog No. 50-002-11398 Supplier Medchemexpress LLC Supplier No. HY10095810MM 1ML
Add to Cart
Add to Cart
4-DAMP (4-diphenylacetoxy-N-methylpiperidinium iodide) is a quaternary ammonium muscarinic receptor antagonist selective for M3 and M5 subtypes, supplied for research applications. It is available as a ready-to-use 10 mM solution in DMSO (1 mL) and in solid forms, and is accompanied by analytical documentation for quality assurance.
  • Ready-to-use 10 mM solution in DMSO for convenient dosing and preparation.
  • High purity suitable for research applications (>98% by HPLC).
  • Appropriate for pharmacological, receptor binding, and signaling studies.
  • Supplied with supporting analytical documents such as COA, HNMR, and RP-HPLC.
  • Molecular weight 451.3 g/mol; CAS 1952-15-4.