Learn More
Medchemexpress LLC 4-(diphenylmethoxy)-1-[3-(1H-tetrazol-5-yl)propyl]-piperidine | 162641-16-9 | MFCD00936179 | >98.0% | 377.48 | C22H27N5O | 100 MG
SDP
Catalog No. 5000242616
Encompass_Preferred
HQL-79 is a potent, selective, and orally active inhibitor of human hematopoietic prostaglandin D synthase (H-PGDS) commonly used in biochemical and cellular research to study prostaglandin D2-mediated signaling and anti-allergic or anti-inflammatory mechanisms.
  • Potent H-PGDS inhibition (IC50 = 6 μM; Kd ≈ 0.8 μM).
  • Selective profile with minimal activity on COX-1, COX-2, m-PGES, and L-PGDS.
  • Oral activity supports in vivo pharmacology studies.
  • High purity suitable for biochemical and cellular assays (>98%).
  • Applicable to allergy and inflammation research focused on PGD2 signaling.
Catalog No. 50-002-42616 Supplier Medchemexpress LLC Supplier No. HY108259100MG
HQL-79 is a potent, selective, and orally active inhibitor of human hematopoietic prostaglandin D synthase (H-PGDS) commonly used in biochemical and cellular research to study prostaglandin D2-mediated signaling and anti-allergic or anti-inflammatory mechanisms.
  • Potent H-PGDS inhibition (IC50 = 6 μM; Kd ≈ 0.8 μM).
  • Selective profile with minimal activity on COX-1, COX-2, m-PGES, and L-PGDS.
  • Oral activity supports in vivo pharmacology studies.
  • High purity suitable for biochemical and cellular assays (>98%).
  • Applicable to allergy and inflammation research focused on PGD2 signaling.