Learn More
Medchemexpress LLC 4-(Diphenylmethoxy)-1-[3-(1H-tetrazol-5-yl)propyl]piperidine | 162641-16-9 | MFCD00936179 | >98.0% | 377.5 g/mol | C22H27N5O | 50 MG
SDP
Catalog No. 5000242678
Encompass_Preferred
HQL-79 (CAS 162641-16-9) is a selective hematopoietic prostaglandin D synthase (H-PGDS) inhibitor supplied for research use. It is orally active in biological assays and inhibits PGD2 synthesis, with reported biochemical potency (IC50 ≈ 6 μM; Kd ≈ 0.8 μM). The compound is supplied as a crystalline solid (C22H27N5O; MW 377.5 g/mol) and is typically stored frozen to preserve integrity.
  • Selective H-PGDS inhibitor for mechanistic studies.
  • Reported IC50 ≈ 6 μM and Kd ≈ 0.8 μM.
  • Orally active in biological assays.
  • High purity (reported >98%).
  • Crystalline solid with formula C22H27N5O and MW 377.5 g/mol.
  • Recommended storage at -20°C to maintain stability.
Catalog No. 50-002-42678 Supplier Medchemexpress LLC Supplier No. HY10825950MG
HQL-79 (CAS 162641-16-9) is a selective hematopoietic prostaglandin D synthase (H-PGDS) inhibitor supplied for research use. It is orally active in biological assays and inhibits PGD2 synthesis, with reported biochemical potency (IC50 ≈ 6 μM; Kd ≈ 0.8 μM). The compound is supplied as a crystalline solid (C22H27N5O; MW 377.5 g/mol) and is typically stored frozen to preserve integrity.
  • Selective H-PGDS inhibitor for mechanistic studies.
  • Reported IC50 ≈ 6 μM and Kd ≈ 0.8 μM.
  • Orally active in biological assays.
  • High purity (reported >98%).
  • Crystalline solid with formula C22H27N5O and MW 377.5 g/mol.
  • Recommended storage at -20°C to maintain stability.