Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(Diphenylmethoxy)-1-[3-(1H-tetrazol-5-yl)propyl]-piperidine | 162641-16-9 | MFCD00936179 | 99.9% | 377.48 | C22H27N5O | 10 MG
SDP

Catalog No. 5000242617
Encompass_Preferred

HQL-79 is a selective, orally active inhibitor of hematopoietic prostaglandin D synthase (H-PGDS) used as a research reagent to investigate prostaglandin D2-related signaling and allergic inflammation. It is supplied as a high-purity solid intended for laboratory research use only.

  • Selective H-PGDS inhibitor for prostaglandin D2 pathway research.
  • High purity: 99.9%.
  • Chemical formula: C22H27N5O; molecular weight: 377.48.
  • Appearance: white to off-white solid.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Provided as a measured research quantity (10 MG).

Catalog No. 50-002-42617 Supplier Medchemexpress LLC Supplier No. HY10825910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HQL-79 is a selective, orally active inhibitor of hematopoietic prostaglandin D synthase (H-PGDS) used as a research reagent to investigate prostaglandin D2-related signaling and allergic inflammation. It is supplied as a high-purity solid intended for laboratory research use only.

  • Selective H-PGDS inhibitor for prostaglandin D2 pathway research.
  • High purity: 99.9%.
  • Chemical formula: C22H27N5O; molecular weight: 377.48.
  • Appearance: white to off-white solid.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Provided as a measured research quantity (10 MG).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.