Learn More
Medchemexpress LLC 4-dodecyl-N-(1,3,4-thiadiazol-2-yl)benzenesulfonamide | 1191951-57-1 | 98.1% | 100 MG
SDP
Catalog No. 5000230782
Encompass_Preferred
PHT-427 is a small-molecule research inhibitor that binds the pleckstrin homology (PH) domain of Akt and inhibits PDPK1. It is used in preclinical signaling studies with reported Kis of 2.7 μM (Akt PH domain) and 5.2 μM (PDPK1); supplied as a white to off-white solid with high purity for laboratory research use only.
  • Inhibits the pleckstrin homology (PH) domain of Akt and PDPK1.
  • Reported Ki values of 2.7 μM (Akt PH domain) and 5.2 μM (PDPK1).
  • High purity (≈98.1%) suitable for research applications.
  • Solid, white to off-white appearance for standard handling.
  • Supplied in defined milligram quantities for dosing and formulation studies.
Catalog No. 50-002-30782 Supplier Medchemexpress LLC Supplier No. HY12063100MG
PHT-427 is a small-molecule research inhibitor that binds the pleckstrin homology (PH) domain of Akt and inhibits PDPK1. It is used in preclinical signaling studies with reported Kis of 2.7 μM (Akt PH domain) and 5.2 μM (PDPK1); supplied as a white to off-white solid with high purity for laboratory research use only.
  • Inhibits the pleckstrin homology (PH) domain of Akt and PDPK1.
  • Reported Ki values of 2.7 μM (Akt PH domain) and 5.2 μM (PDPK1).
  • High purity (≈98.1%) suitable for research applications.
  • Solid, white to off-white appearance for standard handling.
  • Supplied in defined milligram quantities for dosing and formulation studies.