Learn More
Sigma Aldrich 4-Dodecylbenzenesulfonic acid
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 4-dodecylbenzenesulfonic acid (dbsa) is an alkyl sulfonate. it participates in the preparation of a polymer along with polystyrene-block-poly(4-vinylpyridine) and a comb-coil a-block-(b-graft-c) type of copolymer.4-dodecylbenzenesulfonic acid (p-dodecylbenzenesulfonic acid, dbsa) may be used:
- As a solvent for the synthesis of 2-(1,5-diaryl-1,4-pentadien-3-ylidene)-hydrazinecarboximidamide hydrochlorides
- In situ coating of rutile nanoparticles
- As a stabilizer in the preparation of colloidal cds
- In the fabrication of solid-state dye-sensitized solar cells (dsscs)
Specifications
Specifications
| CAS | 121-65-3 |
| Density | 1.06 g/mL (at 20°C (literature)) |
| Molecular Formula | C18H30O3S |
| Linear Formula | C18H30O3S |
| MDL Number | MFCD00066677 |
| Quantity | 1 L |
| Molecular Weight (g/mol) | 326.49 |
| Percent Purity | ≥95% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.