Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Dodecylbenzenesulfonic acid

Catalog No. 44198250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-198-250ML 250 mL
44-198-1L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 44-198-250ML Supplier Sigma Aldrich Supplier No. 44198250ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-dodecylbenzenesulfonic acid (dbsa) is an alkyl sulfonate. it participates in the preparation of a polymer along with polystyrene-block-poly(4-vinylpyridine) and a comb-coil a-block-(b-graft-c) type of copolymer.4-dodecylbenzenesulfonic acid (p-dodecylbenzenesulfonic acid, dbsa) may be used:
  • As a solvent for the synthesis of 2-(1,5-diaryl-1,4-pentadien-3-ylidene)-hydrazinecarboximidamide hydrochlorides
  • In situ coating of rutile nanoparticles
  • As a stabilizer in the preparation of colloidal cds
  • In the fabrication of solid-state dye-sensitized solar cells (dsscs)

Specifications

CAS 121-65-3
Density 1.06 g/mL (at 20°C (literature))
Molecular Formula C18H30O3S
Linear Formula C18H30O3S
MDL Number MFCD00066677
Quantity 250 mL
Molecular Weight (g/mol) 326.49
Percent Purity ≥95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.