Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-dodecylmorpholine | 1541-81-7 | MFCD00082177 | 99.8% | 255.44 g/mol | C16H33NO | 250 MG
SDP

Catalog No. 5000284306
Encompass_Preferred

4-Dodecylmorpholine is a long-chain morpholine surfactant (chemical formula C16H33NO; molecular weight 255.44 g/mol) used as a hydrophilic enzyme detergent, an extraction collector in reverse foam preparation, and in research on antimalarial activity. The reagent is supplied for research use at high reported purity (99.82%, listed as 99.8%) and is commonly offered in small pack sizes such as 250 MG.

  • High purity: 99.8%.
  • Molecular weight: 255.44 g/mol; chemical formula C16H33NO.
  • Common applications: surfactant, hydrophilic enzyme detergent, extraction collector for reverse foam preparation, antimalarial research.
  • Available pack size suitable for research: 250 MG.
  • CAS number 1541-81-7 for unambiguous identification.

Catalog No. 50-002-84306 Supplier Medchemexpress LLC Supplier No. HYW612975250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Dodecylmorpholine is a long-chain morpholine surfactant (chemical formula C16H33NO; molecular weight 255.44 g/mol) used as a hydrophilic enzyme detergent, an extraction collector in reverse foam preparation, and in research on antimalarial activity. The reagent is supplied for research use at high reported purity (99.82%, listed as 99.8%) and is commonly offered in small pack sizes such as 250 MG.

  • High purity: 99.8%.
  • Molecular weight: 255.44 g/mol; chemical formula C16H33NO.
  • Common applications: surfactant, hydrophilic enzyme detergent, extraction collector for reverse foam preparation, antimalarial research.
  • Available pack size suitable for research: 250 MG.
  • CAS number 1541-81-7 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.