Learn More
Fire King Security Group 4 drw, 1 hr fire & impact rated, 31 in wide lateral Parchment Finish
SDP
Supplier:  Fire King Security Group 43122CPA
Encompass_Preferred
Four-drawer lateral 31 in wide UL Class 350 one-hour fire / impact rated filing cabinet with one high-security UL listed keylock top drawer to control all drawers. ETL Verified Mark 1/2 hour fire protection hard drives, CDs, DVDs, flash drives.
Catalog No. NC2952208
Add to Cart
Request a Quote