Learn More
Ace Glass, Inc. 4 dram poly-seal vial, cs/1152, sp/144
SDP
Catalog No. 50976410
Encompass
4 dram sample vials fabricated from borosilicate glass. Include poly-seal cone-lined closures, which are molded in a premium grade, low density polyethylene. Inverted cone liner provides excellent seal and torque retention. Pack/144.
Catalog No. 50-976-410 Supplier Ace Glass, Inc. Supplier No. 877940
4 dram sample vials fabricated from borosilicate glass. Include poly-seal cone-lined closures, which are molded in a premium grade, low density polyethylene. Inverted cone liner provides excellent seal and torque retention. Pack/144.