Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC [4-[(E)-2-(1H-indazol-3-yl)ethenyl]phenyl]-piperazin-1-ylmethanone | 1000669-72-6 | MFCD18385006 | 99.9% | 332.40 g·mol-1 | C20H20N4O | 100 MG
SDP

Catalog No. 5000244548
Encompass_Preferred

KW-2449 is a small-molecule, multi-targeted kinase inhibitor used in research to study signaling pathways and leukemia models. Supplied as a high-purity white powder, it is characterized by the molecular formula C20H20N4O and a molecular weight of 332.40 g·mol-1.

  • Multi-targeted kinase inhibition: inhibits FLT3, ABL, ABL T315I, and Aurora kinases at low nanomolar IC50s.
  • High purity: reported 99.91% purity suitable for biochemical and cell-based assays.
  • Solid powder form: convenient for dissolution in common organic solvents and buffers.
  • Well-characterized: SMILES and molecular properties available for computational and analytical use.
  • For research use only: not intended for human or clinical use.

Catalog No. 50-002-44548 Supplier Medchemexpress LLC Supplier No. HY10339100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KW-2449 is a small-molecule, multi-targeted kinase inhibitor used in research to study signaling pathways and leukemia models. Supplied as a high-purity white powder, it is characterized by the molecular formula C20H20N4O and a molecular weight of 332.40 g·mol-1.

  • Multi-targeted kinase inhibition: inhibits FLT3, ABL, ABL T315I, and Aurora kinases at low nanomolar IC50s.
  • High purity: reported 99.91% purity suitable for biochemical and cell-based assays.
  • Solid powder form: convenient for dissolution in common organic solvents and buffers.
  • Well-characterized: SMILES and molecular properties available for computational and analytical use.
  • For research use only: not intended for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.