Learn More
Medchemexpress LLC {4-[(E)-2-(1H-indazol-3-yl)vinyl]phenyl}(1-piperazinyl)methanone | 1000669-72-6 | MFCD18385006 | 99.9% | 332.40 g·mol⁻¹ | C20H20N4O | 1 ML
SDP
Catalog No. 5000244741
Encompass_Preferred
KW-2449 is a multi-targeted small-molecule kinase inhibitor active against FLT3, ABL (including ABL T315I), and Aurora kinases. It exhibits low-nanomolar potency and is supplied for laboratory research use.
  • Multi-targeted kinase inhibitor effective against FLT3, ABL, and Aurora kinases.
  • Low-nanomolar potency with reported IC50s of 6.6, 14, 4, and 48 nM (FLT3, ABL, ABL T315I, Aurora).
  • Provided as a 10 mM solution in DMSO (1 mL) or as a solid in multiple milligram sizes.
  • High purity suitable for research (reported purity 99.9%).
  • Molecular formula C20H20N4O; molecular weight 332.40 g·mol⁻¹.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-44741 Supplier Medchemexpress LLC Supplier No. HY1033910MM 1ML
KW-2449 is a multi-targeted small-molecule kinase inhibitor active against FLT3, ABL (including ABL T315I), and Aurora kinases. It exhibits low-nanomolar potency and is supplied for laboratory research use.
  • Multi-targeted kinase inhibitor effective against FLT3, ABL, and Aurora kinases.
  • Low-nanomolar potency with reported IC50s of 6.6, 14, 4, and 48 nM (FLT3, ABL, ABL T315I, Aurora).
  • Provided as a 10 mM solution in DMSO (1 mL) or as a solid in multiple milligram sizes.
  • High purity suitable for research (reported purity 99.9%).
  • Molecular formula C20H20N4O; molecular weight 332.40 g·mol⁻¹.
  • For research use only; not for human or veterinary use.