Learn More
Medchemexpress LLC 4-[(E)-2-[4-[(E)-2-(4-hydroxyphenyl)ethenyl]-3-methoxyphenyl]ethenyl]phenol | 863918-78-9 | MFCD25563254 | 98.5% | 344.40 | C23H20O3 | 1 ML
SDP
Catalog No. 5000246253
Encompass_Preferred
Methoxy-X04 is a blood-brain barrier permeant fluorescent dye that selectively binds beta-pleated sheets in dense-core amyloid-β plaques for in vivo and ex vivo imaging and histological staining. It is supplied as a 10 mM solution in DMSO or as a solid for reconstitution.
  • Blood-brain barrier permeant fluorescent probe.
  • Selective binding to amyloid-β plaques, tangles, and cerebrovascular amyloid.
  • Excitation 370 nm; emission 452 nm for blue fluorescence detection.
  • Available as 10 mM solution in DMSO or as solids in multiple pack sizes.
  • High purity (~98.5%) and molecular weight 344.40 Da.
Catalog No. 50-002-46253 Supplier Medchemexpress LLC Supplier No. HY10324010MM 1ML
Add to Cart
Add to Cart
Methoxy-X04 is a blood-brain barrier permeant fluorescent dye that selectively binds beta-pleated sheets in dense-core amyloid-β plaques for in vivo and ex vivo imaging and histological staining. It is supplied as a 10 mM solution in DMSO or as a solid for reconstitution.
  • Blood-brain barrier permeant fluorescent probe.
  • Selective binding to amyloid-β plaques, tangles, and cerebrovascular amyloid.
  • Excitation 370 nm; emission 452 nm for blue fluorescence detection.
  • Available as 10 mM solution in DMSO or as solids in multiple pack sizes.
  • High purity (~98.5%) and molecular weight 344.40 Da.