Learn More
California Pacific Lab & Consulting 4" ECO Funnel with 70mm SW cap adapter (6TPI Buttress)
SDP
Catalog No. NC1361230
Encompass_Preferred
This 4" ECO Funnel is designed to fit a 20 liter (5 gallon) Rectangular Samson Stacker Drum with a 70mm 6TPI closure. Our patented ECO Funnel features a latching lid with gasket to keep fumes contained. A removable large particle filter catches stir bars or other items. A detachable tube extends into the waste container and reduces chemical evaporation.
Catalog No. NC1361230 Supplier California Pacific Lab & Consulting Supplier No. EF4SW
This 4" ECO Funnel is designed to fit a 20 liter (5 gallon) Rectangular Samson Stacker Drum with a 70mm 6TPI closure. Our patented ECO Funnel features a latching lid with gasket to keep fumes contained. A removable large particle filter catches stir bars or other items. A detachable tube extends into the waste container and reduces chemical evaporation.