Learn More
California Pacific Lab & Consulting 4'-ECOFUNNEL W/70MM CAP 1/PK
SDP
Catalog No. NC9536338
Encompass_Preferred
4 in. SAFETY ECO FUNNEL FOR LIQUID WASTE COLLECTION, FITS 70MM SCREW CAP CARBOYS/TIGHT HEAD DRUMS, PREVENTS 99.9% EMISSIONS OF TOXIC FUMES, HDPE, RED LID WITH GASKET THAT LATCHES, REMOVABLE FILTER FOR LARGE PARTICLES, 450 ML CAPACITY, AIDS IN FIRE PREVENTIO
Catalog No. NC9536338 Supplier California Pacific Lab & Consulting Supplier No. EF4FS70
4 in. SAFETY ECO FUNNEL FOR LIQUID WASTE COLLECTION, FITS 70MM SCREW CAP CARBOYS/TIGHT HEAD DRUMS, PREVENTS 99.9% EMISSIONS OF TOXIC FUMES, HDPE, RED LID WITH GASKET THAT LATCHES, REMOVABLE FILTER FOR LARGE PARTICLES, 450 ML CAPACITY, AIDS IN FIRE PREVENTIO