Learn More
California Pacific Lab & Consulting 4 In ECO Funnel With SS Quick Disconnect, Chemically Resistant HDPE, 99.9% Fume Emission Reduction, Each
SDP
Catalog No. NC0885573
Encompass_Preferred
The 4 inch ECO Funnel is designed for safe collection of hazardous liquid waste in lab environments. Featuring a stainless steel quick disconnect adapter, it effectively integrates with Justrite Poly HPLC Waste Disposal Cans. Constructed from HDPE, it boasts a latching lid with an airtight seal, a removable stir-bar catcher, and capability for up to 6 HPLC lines, reducing fume emissions by 99.9%. This ensures enhanced safety and compliance with safety regulations.
Catalog No. NC0885573 Supplier California Pacific Lab & Consulting Supplier No. EF438008N
The 4 inch ECO Funnel is designed for safe collection of hazardous liquid waste in lab environments. Featuring a stainless steel quick disconnect adapter, it effectively integrates with Justrite Poly HPLC Waste Disposal Cans. Constructed from HDPE, it boasts a latching lid with an airtight seal, a removable stir-bar catcher, and capability for up to 6 HPLC lines, reducing fume emissions by 99.9%. This ensures enhanced safety and compliance with safety regulations.