Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-epi-edoxaban tosylate | 2852734-47-3 | 720.26 g/mol | C31H38ClN7O7S2 | 1 MG
SDP

Catalog No. 5000210667
Encompass_Preferred

4-epi-Edoxaban tosylate is the tosylate salt of the 4-epimer of edoxaban, a small-molecule factor Xa inhibitor used as a research compound. Intended for laboratory research use only; not for human or clinical use.

  • Small-molecule factor Xa inhibitor epimer.
  • Available as a solid in milligram quantities for research applications.
  • Cas number 2852734-47-3; molecular weight 720.26 g/mol.
  • Molecular formula C31H38ClN7O7S2.
  • Recommended storage sealed at 4°C; in solution store at -80°C for up to 6 months or -20°C for up to 1 month.
  • For research use only; not for therapeutic use.

Catalog No. 50-002-10667 Supplier Medchemexpress LLC Supplier No. HYW062274A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-epi-Edoxaban tosylate is the tosylate salt of the 4-epimer of edoxaban, a small-molecule factor Xa inhibitor used as a research compound. Intended for laboratory research use only; not for human or clinical use.

  • Small-molecule factor Xa inhibitor epimer.
  • Available as a solid in milligram quantities for research applications.
  • Cas number 2852734-47-3; molecular weight 720.26 g/mol.
  • Molecular formula C31H38ClN7O7S2.
  • Recommended storage sealed at 4°C; in solution store at -80°C for up to 6 months or -20°C for up to 1 month.
  • For research use only; not for therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.