Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals 4-Epitetracycline hydrochloride, can be used as a secondary standard

Catalog No. AC233125000
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC233125000 500 mg Glass bottle
AC233121000 100 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC233125000 Supplier Thermo Scientific Chemicals Supplier No. 233125000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

4-Epitetracycline hydrochloride, CAS # 23313-80-6, is a degradation product of tetracycline that shows antibiotic activity and can induce the protein biosynthesis inhibition.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • 4-Epitetracycline hydrochloride is a metabolite derived from the degradation of tetracycline compounds and has been shown to display antibiotic activity
  • This compound blocks protein synthesis by binding to the 30S ribosomal subunit and interfering with the binding of aminoacyl-tRNA to the mRNA-ribosome complex

Applications

  • This semi-synthesized compound derived from actinomycetes can decrease the activity of the Tet repressor effector in animal models
  • In vivo experiments revealed that 4-epitetracycline hydrochloride can induce a toxic response
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23313-80-6
Molecular Formula C22H24N2O8·HCl
Molecular Weight (g/mol) 480.89
MDL Number MFCD00865028
InChI Key YCIHPQHVWDULOY-DXDJYCPMSA-N
Synonym 4-epitetracycline hydrochloride, epitetracycline hydrochloride, 4-epi-tetracycline hydrochloride, quatrimycin hydrochloride, epitetracycline hcl, unii-i7c3edl2yg, i7c3edl2yg, epitetracycline hydrochloride usp, 4r,4as,5as,6s,12as-4-dimethylamino-1,4,4a,5,5a,6,11,12a-octahydro-3,6,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide monohydrochloride
PubChem CID 54686189
IUPAC Name (4R,4aS,5aS,6S,12aR)-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
SMILES CC1(C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.Cl

Specifications

Melting Point 217.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 80% min. (as is,calculated)
Packaging Glass bottle
Quantity 500 mg
Specific Rotation − 322.00
Solubility Information Solubility in water: 100g/L. Other solubilities: 100g/L methanol
Formula Weight 480.89
Percent Purity ≥97.0% Total Base (on anhydrous substance), ≥80 wt% (as is, calculated) (HPLC)
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Epitetracycline hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Suspected of damaging the unborn child.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECSNumber QJ1898250

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.