Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethoxy-1,3-phenylenediamine Dihydrochloride 98%, Thermo Scientific™

Catalog No. AC190540500
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC190540500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC190540500 Supplier Thermo Scientific Chemicals Supplier No. 190540500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67801-06-3
Molecular Formula C8H14Cl2N2O
Molecular Weight (g/mol) 225.113
InChI Key WDQOMIASNLKYHY-UHFFFAOYSA-N
Synonym unii-3mlj55e9h7, 2,4-diaminophenetole, dihydrochloride, 2,4-diaminoethoxybenzene dihydrochloride, 4-ethoxybenzene-1,3-diammonium dichloride, 4-ethoxy-1,3-phenylenediamine, dihydrochloride, 1,3-benzenediamine, 4-ethoxy-, dihydrochloride, 4-ethoxy-1,3-benzenediamine dihydrochloride, 1,3-benzenediamine, 4-ethoxy-, hydrochloride 1:2, 4-ethoxybenzene-1,3-diamine dihydrochloride, m-phenylenediamine, 4-ethoxy-, dihydrochloride
PubChem CID 49980
IUPAC Name 4-ethoxybenzene-1,3-diamine;dihydrochloride
SMILES CCOC1=C(C=C(C=C1)N)N.Cl.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Gray
Infrared Spectrum Authentic
Quantity 50 g
Formula Weight 225.12
Percent Purity ≥97.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Ethoxy-1, 3-phenylenediamine dihydrochloride, 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.