Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethoxy-2,3-difluoro-4'-(trans-4-propylcyclohexyl)biphenyl 98.0+%, TCI America™
SDP

Catalog No. E11585G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11585G 5 g
E115825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11585G Supplier TCI America Supplier No. E11585G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 189750-98-9
Molecular Formula C23H28F2O
Molecular Weight (g/mol) 358.473
MDL Number MFCD18451991
InChI Key IBFAIOMGVHPWRQ-UHFFFAOYSA-N
PubChem CID 18681376
IUPAC Name 1-ethoxy-2,3-difluoro-4-[4-(4-propylcyclohexyl)phenyl]benzene
SMILES CCCC1CCC(CC1)C2=CC=C(C=C2)C3=C(C(=C(C=C3)OCC)F)F

Specifications

Melting Point 79°C
Color White
Quantity 5 g
Formula Weight 358.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethoxy-2,3-difluoro-4′-(trans-4-propylcyclohexyl)biphenyl
EINECSNumber (4)-1967
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.