Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethoxy-2,3-difluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. E11921G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11921G 1 g
E1192-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11921G Supplier TCI America Supplier No. E11921G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 212386-71-5
Molecular Formula C8H9BF2O3
MDL Number MFCD09258743
InChI Key OBKSFBWOZSQGGC-UHFFFAOYSA-N
PubChem CID 15135235
IUPAC Name (4-ethoxy-2,3-difluorophenyl)boronic acid

Specifications

Melting Point 154°C
Color White
Quantity 1 g
Synonym 2,3-difluoro-4-ethoxyphenylboronic acid, 4-ethoxy-2,3-difluorophenyl boronic acid, 2,3-difluoro-4-ethoxybenzeneboronic acid, boronic acid, 4-ethoxy-2,3-difluorophenyl, pubchem9562, 2,3-difluoro-4-ethoxyphenylboronicacid, acmc-209fik, ksc201g9b, 4-ethoxy-2,3-difluorobenzeneboronic acid
SMILES CCOC1=C(F)C(F)=C(C=C1)B(O)O
Molecular Weight (g/mol) 201.96
Formula Weight 201.96
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethoxy-2,3-difluorophenylboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.