Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethoxy-3-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193971 Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11241G 1 g
E1124-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11241G Supplier TCI America Supplier No. E11241G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 279263-10-4
Molecular Formula C8H10BFO3
Molecular Weight (g/mol) 183.97
MDL Number MFCD04115667
InChI Key ZONJMULNQBNBGU-UHFFFAOYSA-N
PubChem CID 2782821
IUPAC Name (4-ethoxy-3-fluorophenyl)boronic acid
SMILES CCOC1=CC=C(C=C1F)B(O)O

Specifications

Melting Point 113°C
Color White
Quantity 1 g
Synonym 4-ethoxy-3-fluorophenyl boronic acid, 4-ethoxy-3-fluorobenzeneboronic acid, 4-ethoxy-3-fluorophenylboronicacid, 3-fluoro-4-ethoxyphenyl boronic acid, boronic acid, 4-ethoxy-3-fluorophenyl, 4-ethoxy-3-fluoro-phenyl boronic acid, acmc-1cp7m, 4-borono-2-fluorophenetole, 3-fluoro-4-ethoxyphenylboronic acid, 4-ethoxy-3-fluorophenyl boronicacid
Formula Weight 183.97
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethoxy-3-fluorophenylboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.