Learn More
eMolecules​ 104089-17-0 | 4-Ethoxy-4-oxobutylzinc iodide, 0.50 M in THF | Synthonix - Stock307.440 | C6H11IO2Zn | 90.000 | [I-].CCOC(=O)CCC[Zn+] | 100ml | 437236904
Catalog No. 502697785 Shop All eMolecules Inc. Products
Encompass_Preferred
4-Ethoxy-4-oxobutylzinc iodide, 0.50 M in THF | Synthonix - Stock | 104089-17-0307.440 | C6H11IO2Zn | 90.000 | [I-].CCOC(=O)CCC[Zn+] | 100ml | 437236904
Catalog No. 50-269-7785 Supplier eMolecules​ Supplier No. 437236904
4-Ethoxy-4-oxobutylzinc iodide, 0.50 M in THF | Synthonix - Stock | 104089-17-0307.440 | C6H11IO2Zn | 90.000 | [I-].CCOC(=O)CCC[Zn+] | 100ml | 437236904