Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Ethoxybenzenesulfonyl chloride

Catalog No. OTV0003191G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV0003191G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV0003191G Supplier Sigma Aldrich Supplier No. OTV0003191G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Reactant for:
  • Catalytic hydrogenation of acylguanidines
  • Strong binding of alkylguanidinium ions by molecular tweezers for a selective arginine receptor molecule
  • Investigations of drug distribution in the body after oral administration to rats

Specifications

CAS 1132-17-8
Molecular Formula C8H9ClO3S
Linear Formula C8H9ClO3S
Quantity 100 g
MDL Number MFCD03147322
Molecular Weight (g/mol) 220.67
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.