Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethoxybenzoyl chloride, 98%

Catalog No. AC434970050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC434970050 5 g
AC434970010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC434970050 Supplier Thermo Scientific Chemicals Supplier No. 434970050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 16331-46-7 | C9H9ClO2 | 184.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16331-46-7
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.62
InChI Key XLWQUESMILVIPR-UHFFFAOYSA-N
Synonym 4-ethoxybenzoylchloride, p-ethoxybenzoyl chloride, p-ethoxy benzoyl chloride, 4-ethoxy-benzoyl chloride, benzoyl chloride, 4-ethoxy, pubchem10868, acmc-209dpj, 4-ethyloxybenzoyl chloride, para-ethoxybenzoyl chloride, ksc493i8n
PubChem CID 140059
IUPAC Name 4-ethoxybenzoyl chloride
SMILES CCOC1=CC=C(C=C1)C(=O)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow or White to Yellow
Boiling Point 128°C to 130°C (6.0 Torr)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Formula Weight 184.62
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 4-Ethoxybenzoyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.