Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethoxybenzoyl Chloride 98.0+%, TCI America™
SDP

Catalog No. E07005G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07005G 5 g
E070025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07005G Supplier TCI America Supplier No. E07005G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16331-46-7
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00000688
InChI Key XLWQUESMILVIPR-UHFFFAOYSA-N
Synonym 4-ethoxybenzoylchloride, p-ethoxybenzoyl chloride, p-ethoxy benzoyl chloride, 4-ethoxy-benzoyl chloride, benzoyl chloride, 4-ethoxy, pubchem10868, acmc-209dpj, 4-ethyloxybenzoyl chloride, para-ethoxybenzoyl chloride, ksc493i8n
PubChem CID 140059
IUPAC Name 4-ethoxybenzoyl chloride
SMILES CCOC1=CC=C(C=C1)C(=O)Cl

Specifications

Color White-Yellow
Boiling Point 145°C
Quantity 5 g
UN Number 3261
Formula Weight 184.62
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Ethoxybenzoyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.