Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Ethoxycoumarin | 35817-27-7 | 99.93% | 190.20 | 5 G
SDP

Catalog No. 5000350119
Encompass_Preferred

4-Ethoxycoumarin is an antimicrobial compound used for research purposes. It has a molecular weight of 190.20 and a purity of 99.93%. The compound should be stored at room temperature for up to 3 years. When in solvent, it can be stored at -80°C for 2 years or -20°C for 1 year. For shipping, it can be transported at room temperature if the duration is less than 2 weeks. Keep the container tightly sealed in a cool, well-ventilated area, away from direct sunlight and sources of ignition.

  • Antimicrobial properties
  • High purity of 99.93%
  • Stable for 3 years at room temperature
  • Stable for 2 years at -80°C in solvent
  • Stable for 1 year at -20°C in solvent
  • Suitable for research use

Catalog No. 50-003-50119 Supplier Medchemexpress LLC Supplier No. HY1385485G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Ethoxycoumarin is an antimicrobial compound used for research purposes. It has a molecular weight of 190.20 and a purity of 99.93%. The compound should be stored at room temperature for up to 3 years. When in solvent, it can be stored at -80°C for 2 years or -20°C for 1 year. For shipping, it can be transported at room temperature if the duration is less than 2 weeks. Keep the container tightly sealed in a cool, well-ventilated area, away from direct sunlight and sources of ignition.

  • Antimicrobial properties
  • High purity of 99.93%
  • Stable for 3 years at room temperature
  • Stable for 2 years at -80°C in solvent
  • Stable for 1 year at -20°C in solvent
  • Suitable for research use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.