Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethoxymethylene-2-phenyl-2-oxazolin-5-one, ≥90% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013246 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3246 1 g
11-101-3248 5 g
11-101-3247 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-3246 Supplier MilliporeSigma Supelco Supplier No. E07531G
Add to Cart
Add to Cart
Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Packaging Poly Bottle
CAS 15646-46-5
Physical Form Neat
Linear Formula C12H11 NO3
MDL Number MFCD00003204
Melting Point 94°C to 96°C
Type Analytical Standard
Industry Type Drug Analysis
Quantity 1 g
Formula Weight 217.22
Percent Purity ≥90% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.