Learn More
4-Ethoxymethylene-2-phenyl-2-oxazolin-5-one, ≥90% (HPLC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| Chemical Name or Material | 4-Ethoxymethylene-2-phenyl-2-oxazolin-5-one |
| Packaging | Poly Bottle |
| CAS | 15646-46-5 |
| Physical Form | Neat |
| Linear Formula | C12H11 NO3 |
| Technique | Drug Analysis |
| MDL Number | MFCD00003204 |
| Melting Point | 94°C to 96°C |
| Type | Analytical Standard |
| Molecular Weight (g/mol) | 217.22 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.