Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethoxymethylene-2-phenyl-2-oxazolin-5-one, 97%, Thermo Scientific™

Catalog No. AC227980100
Change view
Click to view available options
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC227980100 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC227980100 Supplier Thermo Scientific Chemicals Supplier No. 227980100

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 15646-46-5
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C12H11NO3
Quantity 10g
MDL Number MFCD00003204
Synonym oxazolone, oxazolone sensitizing agent, 5 4h-oxazolone, 4-ethoxymethylene-2-phenyl, unii-78il0j7n72, 4-ethoxymethylene-2-phenyl-2-oxazolin-5-one, 4-ethoxymethylene-2-phenyloxazolin-5-one, 4-ethoxymethylene-2-phenyl-2-oxazoline-5-one, 4z-4-ethoxymethylidene-2-phenyl-1,3-oxazol-5-one, 2-oxazolin-5-one, 4-ethoxymethylene-2-phenyl, 4-ethoxymethylene-2-phenyloxazolone
InChI Key SJHPCNCNNSSLPL-NTMALXAHSA-N
SMILES CCOC=C1C(=O)OC(=N1)C2=CC=CC=C2
IUPAC Name (4Z)-4-(ethoxymethylidene)-2-phenyl-1,3-oxazol-5-one
Molecular Weight (g/mol) 217.224
PubChem CID 1712094
Formula Weight 217.22
Percent Purity ≥96.0%
Chemical Name or Material 4-Ethoxymethylene-2-phenyl-2-oxazolin-5-one, 97%
Show More Show Less
EINECSNumber 239-713-9
RTECSNumber RQ5786250
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.