Learn More
Apexbio Technology LLC 4-Ethoxyphenol 622-62-8 500mg
SDP
Catalog No. 5000094545
Encompass_Preferred
4-Ethoxyphenol (CAS 622-62-8) is a phenolic compound isolated from the marine alga-associated bacterium Bacillus amyloliquefaciens SCSIO 00856 In vitro studies have demonstrated that 4-Ethoxyphenol exhibits notable larvicidal activity against amphipod Bacillus larvae with an EC50 value of 24 1 g/mL suggesting interference with essential biological processes in the targeted organisms This molecule is utilized as a tool compound for investigating the mechanisms underlying larval toxicity and holds potential for evaluating its effects in disease models relevant to larval development and survival
Catalog No. 50-000-94545 Supplier Apexbio Technology LLC Supplier No. C6886500
4-Ethoxyphenol (CAS 622-62-8) is a phenolic compound isolated from the marine alga-associated bacterium Bacillus amyloliquefaciens SCSIO 00856 In vitro studies have demonstrated that 4-Ethoxyphenol exhibits notable larvicidal activity against amphipod Bacillus larvae with an EC50 value of 24 1 g/mL suggesting interference with essential biological processes in the targeted organisms This molecule is utilized as a tool compound for investigating the mechanisms underlying larval toxicity and holds potential for evaluating its effects in disease models relevant to larval development and survival