Learn More
Medchemexpress LLC 4-ethoxyphenol | 622-62-8 | MFCD00002334 | 99.0% | 138.16 g/mol | C8H10O2 | 1 ML
SDP
Catalog No. 5000300753
Encompass_Preferred
4-Ethoxyphenol (CAS 622-62-8) is a small phenolic ether used as a research reagent. It has formula C8H10O2 and molecular weight 138.16 g/mol. Offered as a solid and as a ready-to-use 10 mM solution in DMSO (1 mL), it simplifies preparation of stock solutions for biochemical and microbial assays.
  • Small phenolic compound suitable for research applications.
  • Molecular formula C8H10O2; molecular weight 138.16 g/mol.
  • Available as solid or prepared 10 mM solution in DMSO (1 mL).
  • Ready-to-use solution reduces handling and dissolution time.
  • Compatible with typical organic solvent-based assay workflows.
Catalog No. 50-003-00753 Supplier Medchemexpress LLC Supplier No. HYW01578610MM/1ML
4-Ethoxyphenol (CAS 622-62-8) is a small phenolic ether used as a research reagent. It has formula C8H10O2 and molecular weight 138.16 g/mol. Offered as a solid and as a ready-to-use 10 mM solution in DMSO (1 mL), it simplifies preparation of stock solutions for biochemical and microbial assays.
  • Small phenolic compound suitable for research applications.
  • Molecular formula C8H10O2; molecular weight 138.16 g/mol.
  • Available as solid or prepared 10 mM solution in DMSO (1 mL).
  • Ready-to-use solution reduces handling and dissolution time.
  • Compatible with typical organic solvent-based assay workflows.